Merck
  • Selenoprotein W enhances skeletal muscle differentiation by inhibiting TAZ binding to 14-3-3 protein.
Selenoprotein W enhances skeletal muscle differentiation by inhibiting TAZ binding to 14-3-3 protein.
Biochimica et Biophysica Acta (2014)
Jeon YH, Park YH, Lee JH, et al.