Merck
  • Targeting ribosomal S6 kinases/Y-box binding protein-1 signaling improves cellular sensitivity to taxane in prostate cancer.
Targeting ribosomal S6 kinases/Y-box binding protein-1 signaling improves cellular sensitivity to taxane in prostate cancer.
The Prostate (2014-04-15)
Masaki Shiota, Momoe Itsumi, Akira Yokomizo, Ario Takeuchi, Kenjiro Imada, Eiji Kashiwagi, Junichi Inokuchi, Katsunori Tatsugami, Takeshi Uchiumi, Seiji Naito
ZUSAMMENFASSUNG
Taxanes are the only cytotoxic chemotherapeutic agents proved to prolong the survival in patients with castration-resistant prostate cancer. However, because of intrinsic and acquired resistances to taxanes, their therapeutical efficiencies are modest, bringing only a few months of survival benefit. Y-box binding protein-1 (YB-1) promotes cancer cell resistance to various anticancer treatments, including taxanes. Here, we aimed to elucidate the mechanism of taxane resistance by YB-1 and examined overcoming resistance by targeting YB-1 signaling. Gene and protein expression levels were evaluated by quantitative real-time polymerase chain reaction and Western blot analysis, respectively. We evaluated the sensitivity of prostate cancer cells to taxanes using cytotoxicity assays. Natural taxane paclitaxel from Taxus brevifolia activated the Raf-1/extracellular signal-regulated kinase (ERK) pathway, leading to an activation of ribosomal S6 kinases (RSK)/YB-1 signaling. Activated Raf-1/ERK pathway was blunted by YB-1 knockdown in prostate cancer cells, indicating regulation between Raf-1/ERK signaling and YB-1. In addition, ERK or RSK was activated in taxane-resistant prostate cancer cells, resulting in YB-1 activation. YB-1 knockdown as well as RSK inhibition using RSK-specific siRNA or the small molecule inhibitor SL0101 successfully blocked activation of YB-1, leading to suppression of prostate cancer growth and sensitization to paclitaxel. Taken together, these findings indicate that RSK/YB-1 signaling contributes to taxane resistance, and implicate the therapeutics targeting RSK/YB-1 signaling such as RSK inhibitor as a promising novel therapy against prostate cancer, especially in combination with taxane.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Monoklonaler Anti-β-Aktin−Peroxidase-Antikörper der Maus in Maus hergestellte Antikörper, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Paclitaxel, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, natürlich, für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, halbsynthetisch, für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, halbsynthetisch, für die Systemeignung, European Pharmacopoeia (EP) Reference Standard