Merck
  • Effect of cadmium on selected physiological and morphological parameters in metallicolous and non-metallicolous populations of Echium vulgare L.
Effect of cadmium on selected physiological and morphological parameters in metallicolous and non-metallicolous populations of Echium vulgare L.
Ecotoxicology and environmental safety (2014-04-16)
Sławomir Dresler, Wiesław Bednarek, Małgorzata Wójcik
ZUSAMMENFASSUNG
Cadmium tolerance of three populations of Echium vulgare L., naturally occurring on two Zn-Pb waste deposits (metallicolous populations M1, M2) and on an uncontaminated site (non-metallicolous population, NM) was investigated. The plants were cultivated in hydroponics at 0, 5, 15, 30, or 50μM Cd for 14 days. Although Cd reduced the content of photosynthetic pigments indifferently in the three populations, plant growth parameters and root viability analyses confirmed different Cd tolerances decreasing in the order M1>M2>NM in the populations studied. Organic acids (tartrate, malate, citrate, succinate) were not responsible for the elevated Cd tolerance of the metallicolous populations, although malate and citrate might participate in Cd detoxification in the roots of the M1 and M2. Phytochelatin concentrations were higher in the roots of M1 and M2 populations of E. vulgare, suggesting their role in Cd detoxification and different Cd tolerances.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Glutathion reduziert, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathion reduziert, ≥98.0%
Supelco
Glutathion, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Glutathion reduziert, BioXtra, ≥98.0%
Sigma-Aldrich
Cadmium, granular, 30-80 mesh, ≥99%
Sigma-Aldrich
Cadmium, powder, −100 mesh, 99.5% trace metals basis
Sigma-Aldrich
Cadmium, granular, ≥99%, 5-20 mesh
Glutathion, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cadmium, shot, 3 mm, 99.999% trace metals basis
Cadmium, foil, 100x100mm, thickness 1.0mm, as rolled, 99.99%
Cadmium, foil, 10mm disks, thickness 1.0mm, as rolled, 100%
Cadmium, rod, 100mm, diameter 40mm, 99.9%
Cadmium, rod, 25mm, diameter 40mm, 99.9%
Cadmium, rod, 100mm, diameter 10.0mm, 99.999%
Cadmium, foil, 100x100mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 2m coil, thickness 0.5mm, as rolled, 100%
Cadmium, wire reel, 1m, diameter 1.2mm, as drawn, 99.99+%
Cadmium, rod, 200mm, diameter 10.0mm, 99.999%
Cadmium, rod, 50mm, diameter 10.0mm, 99.999%
Cadmium, foil, 25x25mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 25x25mm, thickness 1.0mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 150x150mm, thickness 0.5mm, as rolled, 99.99%
Cadmium, foil, 30x50mm, thickness 0.5mm, as rolled, 99.999%
Cadmium, foil, 50x50mm, thickness 0.125mm, as rolled, 99.99+%
Cadmium, tube, 500mm, outside diameter 2.29mm, inside diameter 1.27mm, wall thickness 0.51mm, 99.95+%
Cadmium, foil, 100x300mm, thickness 0.125mm, as rolled, 99.95%
Cadmium, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cadmium, foil, 50x50mm, thickness 0.25mm, as rolled, 99.99+%
Cadmium, foil, 50x50mm, thickness 2.0mm, as rolled, 99.99%