Merck
Anticoagulation in atrial fibrillation.
BMJ (Clinical research ed.) (2014-04-16)
Benjamin A Steinberg, Jonathan P Piccini
ZUSAMMENFASSUNG
Atrial fibrillation increases the risk of stroke, which is a leading cause of death and disability worldwide. The use of oral anticoagulation in patients with atrial fibrillation at moderate or high risk of stroke, estimated by established criteria, improves outcomes. However, to ensure that the benefits exceed the risks of bleeding, appropriate patient selection is essential. Vitamin K antagonism has been the mainstay of treatment; however, newer drugs with novel mechanisms are also available. These novel oral anticoagulants (direct thrombin inhibitors and factor Xa inhibitors) obviate many of warfarin's shortcomings, and they have demonstrated safety and efficacy in large randomized trials of patients with non-valvular atrial fibrillation. However, the management of patients taking warfarin or novel agents remains a clinical challenge. There are several important considerations when selecting anticoagulant therapy for patients with atrial fibrillation. This review will discuss the rationale for anticoagulation in patients with atrial fibrillation; risk stratification for treatment; available agents; the appropriate implementation of these agents; and additional, specific clinical considerations for treatment.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Alanin, 99%
Supelco
Warfarin, PESTANAL®, analytical standard
Supelco
Warfarin, analytical standard
Sigma-Aldrich
β-Alanin, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
β-Alanin, BioXtra, ≥99.0% (NT)
Supelco
β-Alanin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
β-Alanin, analytical standard
Sigma-Aldrich
β-Alanin, BioUltra, ≥99.0% (NT)