Merck
  • A novel method for tinidazole detection using Mn-modified CdSe/CdS quantum dots as a luminescent probe.
A novel method for tinidazole detection using Mn-modified CdSe/CdS quantum dots as a luminescent probe.
Journal of nanoscience and nanotechnology (2014-04-17)
Yanzi Wu, Yan Lin, Fangdi Wei, Miao Hu, Liping Liu, Ping Zhou, Qin Hu
ZUSAMMENFASSUNG
A new method for the determination of tinidazole based on the fluorescence quenching of citrate-capped Mn-modified CdSe/CdS quantum dots was developed. In aqueous solution, the fluorescence of the quantum dots at 610 nm was quenched gradually with the increase of the concentration of tinidazole. Based on this, a simple, fast, low-cost and specific quantitative method for tinidazole detection was set up. Under optimal conditions, a good linearity was built between the fluorescence quenching of Mn-modified CdSe/CdS quantum dots and the concentration of tinidazole in the range of 4-400 microM with a correlation coefficient of 0.9998. The limit of detection (3sigma/K) was 0.4 microM. The proposed method was applied to the detections of tinidazole in tablets and injections with satisfactory results.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Cadmiumselenid, -325 Mesh particle size, 99.99% trace metals basis, electronic grade
Supelco
Tinidazol, VETRANAL®, analytical standard