Merck
  • Benzene homologues in environmental matrixes from a pesticide chemical region in China: Occurrence, health risk and management.
Benzene homologues in environmental matrixes from a pesticide chemical region in China: Occurrence, health risk and management.
Ecotoxicology and environmental safety (2014-04-17)
Tieyu Wang, Pang Bo, Tan Bing, Zhu Zhaoyun, Du Liyu, Lu Yonglong
ZUSAMMENFASSUNG
The contamination status and health risks of benzene, toluene, ethylbenzene and xylene (BTEX) in air, soil, dust and groundwater were evaluated in a pesticide chemical region located in Hebei province, China. The concentrations of BTEX in air ranged from 7.80 to 238ug/m(3) and those in soil and dust ranged from lower than limit of detection (LOD) to 32,360ng/g dw, and those in groundwater varied from 2.68 to 98.6ug/L. Generally, the levels of BTEX in multimedia matrixes were all below the standards established in China. Health risk assessment was performed based on the monitoring data via inhalation, dermal contact and ingestion pathways and hazard quotient (HQ) was calculated to be on the order of 10(-7), below 10(-6), and Hazard index (HI) levels of BTEX were lower than 1.0. However, both HQ and HI ascended with an increase in work experience/exposure. Integrated risk management was proposed to eliminate BTEX pollution and to protect occupational health of workers in those industries.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Toluol, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluol, anhydrous, 99.8%
Sigma-Aldrich
Benzol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Benzol, ACS reagent, ≥99.0%
Supelco
Benzol, analytical standard
Sigma-Aldrich
Benzol, anhydrous, 99.8%
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzol, puriss. p.a., reag. Ph. Eur., ≥99.7%
Sigma-Aldrich
Ethylbenzol, ReagentPlus®, 99%
Supelco
Benzol -Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol
Supelco
Benzol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol, analytical standard
Sigma-Aldrich
Ethylbenzol, anhydrous, 99.8%
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethylbenzol, analytical standard
Supelco
Toluol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Toluol -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Toluol, ACS reagent, ≥99.5%
Supelco
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in.
Supelco
Toluol-Lösung, certified reference material, TraceCERT®, 200 μg/mL in methanol, ampule of 1 mL
Supelco
Ethylbenzol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D)
Supelco
Ethylbenzol -Lösung, suitable for NMR (reference standard), 10% in chloroform-d (99.8 atom % D), NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.1 %
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d ("100%", 99.96 atom % D)
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 5 mm × 8 in. , ultra-thin wall
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 0.1% in chloroform-d (99.8 atom % D), TMS 0.01 %, NMR tube size 10 mm × 8 in.
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 1% in chloroform-d (99.8 atom % D)
Sigma-Aldrich
Ethylbenzol -Lösung, suitable for NMR (reference standard), 5% in chloroform-d (99.8 atom % D), TMS 2 %, NMR tube size 5 mm × 8 in.