Merck
Controllable synthesis of Cu2O/Cu composites with stable photocatalytic properties.
Journal of nanoscience and nanotechnology (2014-04-18)
Xueqin Liu, Fei Li, Huifang Wang, Jianbo Yang, Zhen Li, Yongqian Wang, Hongyun Jin
ZUSAMMENFASSUNG
Here we reported a facile approach to synthesize Cu2O/Cu composite particles by one-step sono-chemical process. The content of Cu in the Cu2O/Cu composites can be easily controlled by adjusting the synthesis time. Phase, morphology and optical properties of the products were carried out by X-ray diffraction (XRD), field-emission scanning electron microscopy (FSEM), transmission electron microscope (TEM), ultraviolet-visible (UV-vis) spectroscopy and nitrogen adsorption apparatus. Using photocatalytic degradation of Methyl orange (MeO) dye under visible-light illumination, we have investigated the influence of Cu on the photocatalytic activity of Cu2O, to find out its potential application in waste water treatment. Especially, the stability of the photocatalyst was confirmed using reclaimed Cu2O/Cu in ten successive runs. Results demonstrated clearly that Cu2O/Cu were stable and resistant to photocorrosion during the photocatalytic oxidation of organic compounds, indicating that these Cu2O/Cu composites are promising candidates for pollutant processing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kupfer(II)-oxid, ACS reagent, ≥99.0%
Sigma-Aldrich
Kupfer(II)-oxid, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Methylorange, ACS reagent, Dye content 85 %
Sigma-Aldrich
Kupfer(II)-oxid, powder, <10 μm, 98%
Sigma-Aldrich
Kupfer, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer, powder, 99.99% trace metals basis
Sigma-Aldrich
Methylorange, suitable for microscopy
Sigma-Aldrich
Kupfer, powder, <75 μm, 99%
Sigma-Aldrich
Kupfer, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Kupfer, nanopowder, 40-60 nm particle size (SAXS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer(II)-oxid, powder, 99.99% trace metals basis
Sigma-Aldrich
Kupfer, powder (spheroidal), 10-25 μm, 98%
Sigma-Aldrich
Kupfer, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Kupfer, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer(II)-oxid, needles, mixture of CuO and Cu2O, ACS reagent
Sigma-Aldrich
Kupfer, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Kupfer, nanopowder, 60-80 nm particle size (APS), ≥99.5% trace metals basis
Sigma-Aldrich
Kupfer, ACS reagent, granular, 10-40 mesh, ≥99.90%
Sigma-Aldrich
Kupfer, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Kupfer, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Kupfer, foil, thickness 1.0 mm, 99.999% trace metals basis
Kupfer, mesh, 100x100mm, nominal aperture 0.14mm, thickness 0.25mm, wire diameter 0.115mm, 100x100 wires/inch, open area 30.3%, plain weave mesh
Sigma-Aldrich
Kupfer, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Kupfer(II)-oxid, 99.999% trace metals basis
Sigma-Aldrich
Kupfer, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Kupfer, shot, −3-+14 mesh, 99%
Sigma-Aldrich
Kupfer, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Kupfer(II)-oxid, powder, 99.995% trace metals basis
Kupfer, wire reel, 100m, diameter 0.05mm, annealed, 99.9%