Merck
  • ZnO nanocrystals anchored graphene: in situ solvothermal synthesis and enhanced photocatalytic performance.
ZnO nanocrystals anchored graphene: in situ solvothermal synthesis and enhanced photocatalytic performance.
Journal of nanoscience and nanotechnology (2014-04-18)
Zimin Dong, Jun Zhang, Weiwei Yuan, Dong Xie, Gaohui Du
ZUSAMMENFASSUNG
ZnO nanocrystals anchored graphene composite (ZnO/G) was synthesized by a facile in situ solvothermal route. The nanocomposite was characterized by X-ray diffraction (XRD), scanning electron spectroscopy (SEM), transmission electron microscopy (TEM) and Raman spectra (RS). The results indicated that the ZnO particles with an average diameter of ca. 12.6 nm were well-dispersed on the surface of the graphene nanosheets. The optical properties were investigated by fluorescence (PL) and UV diffuse reflectance (UV-VIS). It showed that the nanocomposite displayed a fluorescence quenching property. Furthermore, the nanocomposite showed a remarkably enhanced photocatalytic activity to degrade organic pollutants (MB, MO, Rh-B) under visible light irradiation, with a percentage degradation of MB reaching as high as 99.8% in 60 min.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Zinkoxid, ReagentPlus®, powder, <5 μm particle size, 99.9%
Sigma-Aldrich
Graphit, powder, <20 μm, synthetic
Sigma-Aldrich
Zinkoxid, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Zinkoxid, nanopowder, ≤50 nm particle size
Sigma-Aldrich
Zinkoxid, nanopowder, <50 nm particle size (BET), >97%
Sigma-Aldrich
Zinkoxid, Dispersion, nanoparticles, <100 nm particle size (TEM), ≤40 nm avg. part. size (APS), 20 wt. % in H2O
Sigma-Aldrich
Zinkoxid, puriss., meets analytical specification of Ph. Eur., BP, USP, 99-100.5% (calc. for dried substance)
Sigma-Aldrich
Graphit, flakes
Sigma-Aldrich
Graphit, powder, <45 μm, ≥99.99% trace metals basis
Sigma-Aldrich
Zinkoxid, 99.99% trace metals basis
Sigma-Aldrich
Zinkoxid, 99.999% trace metals basis
Sigma-Aldrich
Zinkoxid, nanowires, Length 1 um
Sigma-Aldrich
Graphit, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Graphit, rod, L 150 mm, diam. 3 mm, low density, 99.995% trace metals basis
Sigma-Aldrich
Graphit, rod, L 150 mm, diam. 6 mm, 99.995% trace metals basis
Supelco
Zinkoxid, analytical standard