Merck
Photo-controllable molecular hydrogels for drug delivery.
Journal of nanoscience and nanotechnology (2014-04-25)
Hanxia Liu, Zhijian Song, Xuemei Chen
ZUSAMMENFASSUNG
A photocleavable nitrobenzyl ester group was introduced in the backbone of self-assembling peptides using standard Fmoc-peptide systhesis. The nitrobenzyl ester group could be used as a cleavable linker to connect a gelator and a hydrophilic group. Photo-irradiation would lead to the generation of the molecular gelator and the formation of a molecular hydrogel. The nitrobenzyl ester group could also be used as one part of hydrophobic capping group for short peptides to produce molecular hydrogelators. In this case, photo-irradiation would lead to the release of hydrophobic anti-imflammatory drug of Naproxen and the gel-sol phase transition. The application of photocleavable group in molecular self-assembly would lead to photo-controllable self-assembly materials for controlled drug release, tissue engineering, and regenerative medicine.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Naproxen, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Naproxen, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Naproxen, meets USP testing specifications
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthalinessigsäure, 98%
Supelco
Naproxen, VETRANAL®, analytical standard
Supelco
Naproxen -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Naproxen, European Pharmacopoeia (EP) Reference Standard