Merck
  • Effect of a proteasome inhibitor plus steroids on HLA antibodies in sensitized patients awaiting a renal transplant.
Effect of a proteasome inhibitor plus steroids on HLA antibodies in sensitized patients awaiting a renal transplant.
Transplantation (2014-04-25)
Olivier Aubert, Caroline Suberbielle, Rémi Gauthe, Helene Francois, Erika Nnang Obada, Antoine Durrbach
ZUSAMMENFASSUNG
This prospective monocentric study investigated the effect of corticosteroids plus bortezomib to reduce anti-HLA antibodies before renal transplantation. Included were 23 patients with stable immunization against HLA and awaiting a kidney transplant (KT). Treatment consisted of bortezomib (1.3 mg/m) plus 40 mg of dexamethasone intravenously on days 1, 3, 8, and 10 (B+S). Class I and II anti-HLAs were determined using the single-antigen beads assay at day 0 (D0), month 1 (M1), M3, and M6. Antibodies against 96 class I and 76 class II antigens were investigated and patients had a mean number of 49 (± 21) antibodies against HLA on D0: 31 were against HLA class I and 17 were against HLA class II. At D0, the median was 10,734 (range, 1096-18,513) for the highest mean fluorescent intensity (hAb) anti-class I antibodies and 11,189 (range, 1276-19,176) for class II. By M3, 41% of patients had a greater than 25% decrease in class 1 hAbs and 60% by M6. By M3, 33% of patients had a greater than 25% decrease in class II hAbs and 42% by M6. At M6, 54% of anti-HLA antibodies had a sustained decrease by more than 25% and 36% were decreased by more than 50%. No predictive factors for decreased antibodies after bortezomib plus steroid therapy were identified. No serious adverse event was observed. Thereafter, 11 of 23 patients received successful transplants without having experienced acute rejection (follow-up, 18 months). B+S is an effective alternative therapy for reducing class I and II anti-HLA, regardless of other previous treatments.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dexamethason, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethason – wasserlöslich, BioReagent, suitable for cell culture
Sigma-Aldrich
Dexamethason, ≥98% (HPLC), powder
Supelco
Dexamethason, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethason, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dexamethason, meets USP testing specifications
Dexamethason, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethason, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethason, tested according to Ph. Eur.
Dexamethason für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dexamethason für die Systemeignung, European Pharmacopoeia (EP) Reference Standard