Merck
Thyroid surgery as a 23-hour stay procedure.
Annals of the Royal College of Surgeons of England (2014-05-02)
A H Perera, S D Patel, N W Law
ZUSAMMENFASSUNG
The main barriers to short stay thyroidectomy are haemorrhage, bilateral recurrent laryngeal nerve palsy causing respiratory compromise and hypocalcaemia. This study assessed the safety and effectiveness of thyroidectomy as a 23-hour stay procedure. All patients undergoing total or completion thyroidectomy were prescribed calcium and vitamin D3 supplements following surgery. Retrospective analysis identified patients admitted for longer than 23 hours and any readmissions. A total of 164 patients were admitted for 23-hour stay thyroid surgery over a 25-month period between 2008 and 2010. Four patients (2%) required admission for longer than 23 hours. No patients required emergency intervention for postoperative haemorrhage or airway compromise. Biochemical hypocalcaemia (despite calcium supplements) was detected in one patient when measured at the outpatient clinic two weeks following surgery. Twelve patients (7.3%) attended the accident and emergency department following discharge; four required admission for intravenous antibiotics for wound infection and one for biochemical hypocalcaemia. This single centre UK experience demonstrates that thyroidectomy can be carried out both safely and effectively as a 23-hour stay procedure. Prophylactic prescription of calcium and vitamin D3 reduces hypocalcaemia, and thereby also prolonged admission and readmission due to hypocalcaemia. Supplements are an acceptable, cost effective method of reducing hypocalcaemia and shortening postoperative length of stay.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Cholecalciferol (Vitamin D3), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Cholecalciferol, ≥98% (HPLC)
Sigma-Aldrich
Calcium, granular, 99%
Supelco
Cholecalciferol (D3), analytical standard
USP
Cholecalciferol, United States Pharmacopeia (USP) Reference Standard
Supelco
Vitamin D3 solution, 1 mg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Cholecalciferol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Cholecalciferol, meets USP testing specifications
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Calcium, pieces, <1 cm, 99%
Sigma-Aldrich
Cholecalciferol, analytical standard
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.9% trace metals basis