Merck
Gabapentin for the prophylaxis of episodic migraine in adults.
American family physician (2014-05-03)
Corey D Fogleman
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
4-Aminobutansäure, ≥99%
Supelco
4-Aminobutansäure, analytical standard
USP
Gabapentin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
4-Aminobutansäure, BioXtra, ≥99%
Supelco
Gabapentin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Gabapentin -Lösung, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Gabapentin, solid
Supelco
4,4-Pentamethylen-2-Pyrrolidinon, Pharmaceutical Secondary Standard; Certified Reference Material
Vigabatrin Unreinheit D, European Pharmacopoeia (EP) Reference Standard