Merck
[Abiraterone acetate (AA): current guidelines of prescription of abiraterone].
Bulletin du cancer (2014-05-06)
Emilie Boissier, Yohann Loriot, Stéphane Vignot, Christophe Massard
ZUSAMMENFASSUNG
Abiraterone acetate (AA) is a selective inhibitor of cytochrom p450 (CYP)17 which is required for androgen biosynthesis, and can block the androgens synthesis by testicles, surrenals and intratumoral secretion. In phase I and II studies in patients with prostate cancer, therapy with AA 250-2000  mg once daily demonstrated reductions in prostate specific antigen (PSA), and/or circulating tumor cells (CTCs). In two large phase III trials in patients with metastatic castration resistant prostate cancer (CRPC) in post-docetaxel and pre-docetaxel setting, AA plus prednisone compared with placebo plus prednisone demonstrated a significant superior overall survival in post-docetaxel setting, and a superior radiological PFS in pre-docetaxel setting. Based of these results, AA is approved in metastatic CRPC patients in post-docetaxel setting or pre-docetaxel setting in 2013.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
USP
Prednison, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Prednison, ≥98%
Supelco
Prednison, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Docetaxel, purum, ≥97.0% (HPLC)
Wasserfreies Docetaxel, European Pharmacopoeia (EP) Reference Standard