Merck
  • Preparation and characterization of epoxy composites filled with functionalized nanosilica particles obtained via sol-gel process
Preparation and characterization of epoxy composites filled with functionalized nanosilica particles obtained via sol-gel process
Polymer (2000)
Kang, S..;