- Dendronized supramolecular nanocapsules: pH independent, water-soluble, deep-cavity cavitands assemble via the hydrophobic effect.
Dendronized supramolecular nanocapsules: pH independent, water-soluble, deep-cavity cavitands assemble via the hydrophobic effect.
Journal of the American Chemical Society (2008-10-14)
Marco D Giles, Simin Liu, Roy L Emanuel, Bruce C Gibb, Scott M Grayson
PMID18847205
ZUSAMMENFASSUNG
At neutral pH, dendronized deep-cavity cavitands were shown to form supramolecular nanocapsules via assembly around a range of guest molecules.