Merck
Stress and water balance: the roles of ANP, AVP and isatin.
Indian journal of experimental biology (1999-03-27)
S K Bhattacharya, A Chakrabarti, V Glover
ZUSAMMENFASSUNG
Stress is often associated with water retention and its resolution with diuresis. The biological systems for the control of stress and water balance are very closely related. Corticotrophin releasing hormone (CRH) and arginine vasopressin (AVP) are co-localised in the hypothalamus and often act synergistically. Atrial natriuretic peptide (ANP) can exert a feedback control on the hypothalamic/pituitary/adrenal axis. ANP has been shown to be anxiolytic, whereas AVP may be anxiogenic. AVP and ANP levels have been found to be abnormal in a range of stress disorders and psychiatric illnesses. Isatin is an endogenous anxiogenic factor which is also a potent inhibitor of the ANP receptor. It may provide a link between the function of monoamines during stress, and the control of water balance by ANP.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Isatin, 97%