- Filgrastim treatment of HIV-infected patients improves neutrophil function.
Filgrastim treatment of HIV-infected patients improves neutrophil function.
AIDS (London, England) (1999-12-22)
D L Pitrak
PMID10596678
ZUSAMMENFASSUNG
The effect of Filgrastim (recombinant methionyl human granulocyte colony-stimulating factor) treatment on neutrophil function was studied in HIV-infected patients. Filgrastim therapy significantly increased oxidative capacity of neutrophils, increased bacterial killing, and reduced accelerated apoptosis of neutrophils observed in this patient population.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Filgrastim, European Pharmacopoeia (EP) Reference Standard