- A systematic review of the clinical efficacy and effectiveness of the holmium:YAG laser in urology.
A systematic review of the clinical efficacy and effectiveness of the holmium:YAG laser in urology.
BJU international (1999-08-12)
I Larizgoitia, J M Pons
PMID10444114
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Holmium, powder, 10g, max. particle size 500 micron, 99.9%