Merck
Urologic applications of the holmium laser.
Urologic nursing (2000-07-13)
G S Young, R Ball
ZUSAMMENFASSUNG
The holmium laser has various urologic applications. It is important for urologic nurses and associates to be familiar with the laser physics and safety recommendations of the holmium laser, as well as to understand the applications and surgical techniques associated with this therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Holmium, chips, 99.9% trace metals basis
Holmium, powder, 10g, max. particle size 500 micron, 99.9%