- Piracetam for fetal distress in labour.
Piracetam for fetal distress in labour.
Piracetam is thought to promote the metabolism of brain cells when they are hypoxic. It has been used to prevent adverse effects of fetal distress. The objective of this review was to assess the effects of piracetam for suspected fetal distress in labour on method of delivery and perinatal morbidity. The Cochrane Pregnancy and Childbirth Group trials register and the Cochrane Controlled Trials Register were searched. Date of last search: February 1999. Randomised trials of piracetam compared with placebo or no treatment for suspected fetal distress in labour. Both reviewers assessed eligibility and trial quality. One study of 96 women was included. Piracetam compared with placebo was associated with a trend to reduced need for caesarean section (relative risk 0.57, 95% confidence interval 0.32 to 1.03). There were no statistically significant differences in relative risk between the piracetam and placebo group for neonatal morbidity (measured by neonatal respiratory distress) or Apgar score. There is not enough evidence to evaluate the use of piracetam for fetal distress in labour.