Merck
Microbial formation, biotechnological production and applications of 1,2-propanediol.
Applied microbiology and biotechnology (2001-03-10)
G N Bennett, K Y San
ZUSAMMENFASSUNG
This short review covers metabolic pathways, genetics and metabolic engineering of 1,2-propanediol formation in microbes. 1,2-Propanediol production by bacteria and yeasts has been known for many years and two general pathways are recognized. One involves the metabolism of deoxyhexoses, where lactaldehyde is formed during the glycolytic reactions and is then reduced to 1,2-propanediol. The second pathway derives from the formation of methylglyoxal from dihydroxyacetonephosphate and its subsequent reduction to 1,2-propanediol. The enzymes involved in the reduction of methylglyoxal can generate isomers of lactaldehyde or acetol, which can be further reduced by specific reductases, giving chiral 1,2-propanediol as the product. The stereospecificity of the enzymes catalyzing the two reduction steps is important in deriving a complete pathway. Through genetic engineering, appropriate combinations of enzymes have been brought together in Escherichia coli and yeast to generate 1,2-propanediol from glucose. The optimization of these strains may yield microbial processes for the production of this widely used chemical.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Propylenglycol, ≥99.5% (GC), FCC, FG
Sigma-Aldrich
Propylenglykol, meets USP testing specifications
Supelco
Propylenglykol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
1,2-Propandiol, ACS reagent, ≥99.5%
USP
Propylenglycol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
1,2-Propandiol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propandiol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
(S)-(+)-1,2-Propandiol, 96%
Sigma-Aldrich
1,2-Propandiol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Supelco
1,2-Propandiol, analytical standard
Propylenglycol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
1,2-Propandiol, tested according to Ph. Eur.