Merck
Final report on the safety assessment of Methyl Alcohol.
International journal of toxicology (2001-05-19)
S Lanigan
ZUSAMMENFASSUNG
Methyl Alcohol is an aliphatic alcohol with use in a few cosmetic formulations as a solvent and denaturant. Concentrations up to 5% are typically used to denature ethyl alcohol in cosmetic products. Methyl Alcohol is readily absorbed through the skin and from the gastrointestinal and respiratory tracts, is distributed throughout all organs and tissues (in direct relation to the body's water distribution), and is eliminated primarily via the lungs. Undiluted Methyl Alcohol is an ocular and skin irritant. Inhalation studies showed a no-effect level for maternal damage of 10,000 ppm and for teratogenic effects of 5,000 ppm. Overall, Methyl Alcohol is not considered mutagenic. Carcinogenicity data were unavailable. The toxicity of Methyl Alcohol in humans results from the metabolism of the alcohol to formate and formic acid through a formaldehyde intermediate. Formate accumulation causes metabolic acidosis and inhibits cellular respiration. Methyl Alcohol toxicity is time and concentration dependent, and its toxic effect is competitively inhibited with ethyl alcohol. Because of the moderating effect of ethyl alcohol, it was concluded that Methyl Alcohol is safe as used to denature ethyl alcohol used in cosmetic products. No conclusion was reached regarding any other use of Methyl Alcohol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.