Merck
2,5-hexanedione-induced testicular injury.
Annual review of pharmacology and toxicology (2002-12-10)
Kim Boekelheide, Shawna L Fleming, Theresa Allio, Michelle E Embree-Ku, Susan J Hall, Kamin J Johnson, Eun Ji Kwon, Sutchin R Patel, Reza J Rasoulpour, Heidi A Schoenfeld, Stephanie Thompson
ZUSAMMENFASSUNG
Now in its third decade of mechanistic investigation, testicular injury caused by 2,5-hexanedione (2,5-HD) exposure is a well-studied model with a rich database. The development of this model reflects the larger changes that have moved biology from a branch of chemistry into the molecular age. Critically examined in this review is the proposed mechanism for 2,5-HD-induced testicular injury in which germ cell maturation is disrupted owing to alterations in Sertoli cell microtubule-mediated functions. The goal is to evaluate the technical and conceptual approaches used to assess 2,5-HD-induced testicular injury, to highlight unanswered questions, and to identify fruitful avenues of future research.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
2,5-Hexandion, ≥98%