Merck
Dynamics of tumour angiogenesis: effect of razoxane-induced growth rate slowdown.
Clinical & experimental metastasis (2003-04-23)
K Hellmann
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Razoxane, >98% (HPLC)