Merck
[A review of the study on swainsonine--toxic elements of locoweed].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2003-08-02)
Y Shi, M Li, F Xu
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Swainsonin, synthetic
Sigma-Aldrich
Swainsonin, from Metarrhizium anisopliae, ≥98% (TLC)