Merck
  • The tyramine conjugation test as a trait marker for endogenous unipolar depression and a predictor of treatment response.
The tyramine conjugation test as a trait marker for endogenous unipolar depression and a predictor of treatment response.
Molecular aspects of medicine (1992-01-01)
J Jarman
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Tyramin, ≥98.0%
Supelco
Tyramin, analytical standard
Sigma-Aldrich
Tyramin, 98%, FG