Merck
Unintentional ingestion of potassium permanganate.
Pediatric emergency care (2004-04-20)
Thomas B Johnson, David D Cassidy
ZUSAMMENFASSUNG
This case of an unintentional ingestion of an unknown amount of potassium permanganate by a 5-year-old boy, and its sequelae, exemplifies the potential danger of this poison. Due to the wide availability of this agent in over-the-counter preparations and the high potential for serious sequelae, clinicians should be aware of the actions of this agent, as well as the diagnostic and management features associated with it.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%
Sigma-Aldrich
Kaliumpermanganat, ACS reagent, ≥99.0%, low in mercury
Sigma-Aldrich
Kaliumpermanganat, 97%
Sigma-Aldrich
Kaliumpermanganat, meets USP testing specifications