Merck
Clinical pharmacokinetics of trospium chloride.
Clinical pharmacokinetics (2005-06-22)
Oxana Doroshyenko, Alexander Jetter, Karl P Odenthal, Uwe Fuhr
ZUSAMMENFASSUNG
Trospium chloride, a quaternary amine with anticholinergic properties, is used for the treatment of overactive bladder with symptoms of urge urinary incontinence, urgency and urinary frequency. The pharmacokinetics of trospium chloride have been investigated in healthy volunteers, in patients with renal and hepatic impairment, and in those with symptoms of overactive bladder, after oral, intravenous and intravesical administration. After oral administration, absorption of the hydrophilic trospium chloride is slow and incomplete. Peak plasma concentrations (Cmax) of approximately 4 ng/mL are reached 4-5 hours after administration of a 20 mg immediate-release preparation. The mean bioavailability is approximately 10% and decreases by concomitant food intake (to a mean of 26% of the fasting area under the plasma concentration-time curve [AUC]). Trospium chloride displays dose proportional increases in AUC and Cmax after a single dose within the clinically relevant dose range (20-60 mg). The mean volume of distribution is approximately 350-800 L. The drug is minimally (mean approximately 10%) metabolised to spiroalcohol by hydrolysis, is 50% plasma protein bound and does not cross the blood-brain barrier. Urinary excretion of the parent compound plays a major role in the disposition of the drug, with a mean renal clearance of 29 L/h (accounting for approximately 70% of total clearance) and a mean elimination half-life ranging from 10 to 20 hours. Elimination of the drug is slowed in patients with renal insufficiency, and population pharmacokinetic modelling has demonstrated that drug clearance is correlated with serum creatinine concentration. Thus, dose reduction is needed in patients with severe renal impairment (i.e. creatinine clearance < 30 mL/min). To date, no clinically relevant pharmacokinetic drug-drug interactions have been identified; the drug does not bind to any of the drug metabolising cytochrome P450 enzymes. The pharmacokinetics of the drug are compatible with twice-daily administration. A once-daily schedule may also be appropriate, but this regimen needs formal clinical evaluation.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Trospiumchlorid, European Pharmacopoeia (EP) Reference Standard