- An overview of synthetic approaches to porphyrin, phthalocyanine, and phenothiazine photosensitizers for photodynamic therapy.
An overview of synthetic approaches to porphyrin, phthalocyanine, and phenothiazine photosensitizers for photodynamic therapy.
Journal of environmental pathology, toxicology and oncology : official organ of the International Society for Environmental Toxicology and Cancer (2006-03-29)
Stephen A Gorman, Stanley B Brown, John Griffiths
PMID16566711
ZUSAMMENFASSUNG
The mechanistic basis of photodynamic therapy is reviewed briefly, and the factors that are important in improving photosensitizers are examined. The methods for preparing the three main classes of PDT photosensitizers, namely, porphyrins, phthalocyanines, and phenothiazines, are reviewed.