Merck
[Update in asthma therapy. Slow-release theophylline].
Nihon Naika Gakkai zasshi. The Journal of the Japanese Society of Internal Medicine (2006-09-08)
Hironori Sagara
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Theophyllin, anhydrous, ≥99%, powder
Supelco
Theophyllin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Theophyllin, United States Pharmacopeia (USP) Reference Standard
Theophyllin, European Pharmacopoeia (EP) Reference Standard