Merck
  • Allergy to tea tree oil: retrospective review of 41 cases with positive patch tests over 4.5 years.
Allergy to tea tree oil: retrospective review of 41 cases with positive patch tests over 4.5 years.
The Australasian journal of dermatology (2007-05-31)
Tim Rutherford, Rosemary Nixon, Mei Tam, Bruce Tate
ZUSAMMENFASSUNG
Tea tree oil use is increasing, with considerable interest in it being a 'natural' antimicrobial. It is found in many commercially available skin and hair care products in Australia. We retrospectively reviewed our patch test data at the Skin and Cancer Foundation Victoria over a 4.5-year period and identified 41 cases of positive reactions to oxidized tea tree oil of 2320 people patch-tested, giving a prevalence of 1.8%. The tea tree oil reaction was deemed relevant to the presenting dermatitis in 17 of 41 (41%) patients. Of those with positive reactions, 27 of 41 (66%) recalled prior use of tea tree oil and eight of 41 (20%) specified prior application of neat (100%) tea tree oil. Tea tree oil allergic contact dermatitis is under-reported in the literature but is sufficiently common in Australia to warrant inclusion of tea tree oil, at a concentration of 10% in petrolatum, in standard patch-test series. Given tea tree oil from freshly opened tea tree oil products elicits no or weak reactions, oxidized tea tree oil should be used for patch testing.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Teebaumöl, FG