Merck
  • All-trans and 13-cis-retinoic acids are normal constituents of human blood formed from all-trans-retinyl palmitate.
All-trans and 13-cis-retinoic acids are normal constituents of human blood formed from all-trans-retinyl palmitate.
Nutrition reviews (1991-09-01)
ZUSAMMENFASSUNG
All-trans and 13-cis-retinoic acids are normal constituents of human blood. Blood levels of both compounds increase upon oral ingestion of all-trans-retinyl palmitate, to a greater extent with higher amounts of ingested retinyl palmitate. These data suggest a mechanism whereby levels of dietary vitamin A could influence blood levels of the known cancer chemopreventive retinoic acids.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
Retinylpalmitat, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Retinylpalmitat, potency: ≥1,700,000 USP units per g
Supelco
Retinolpalmitat, analytical standard
Sigma-Aldrich
Retinylpalmitat, Type IV, ~1,800,000 USP units/g, oil