Merck
Menthol: a refreshing look at this ancient compound.
Journal of the American Academy of Dermatology (2007-05-15)
Tejesh Patel, Yozo Ishiuji, Gil Yosipovitch
ZUSAMMENFASSUNG
Menthol is a naturally occurring cyclic terpene alcohol of plant origin, which has been used since antiquity for medicinal purposes. Its use in dermatology is ubiquitous, where it is frequently part of topical antipruritic, antiseptic, analgesic, and cooling formulations. Despite its widespread use, it was only recently that the mechanism by which menthol elicits the same cool sensation as low temperature was elucidated upon, with the discovery of the TRPM8 receptor. Although almost 5 years have passed since the discovery of this receptor, many dermatologists are still unaware of menthol's underlying target. The purpose of this review is to highlight the recent advances in the mechanism of action of menthol and to provide an overview of its dermatologic applications.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Supelco
(−)-Menthol, analytical standard
Menthol, European Pharmacopoeia (EP) Reference Standard