Merck
[Advances in research on pharmacodynamics and chemical conversion of catalpol].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2007-09-07)
Yan-fei Liu, Yu Zhao, Xue-sen Wen, Qi-ting Dong
ZUSAMMENFASSUNG
Catalpol is a active component in Rehmannia Root, and its pharmacological effects are extensive. In this paper its effects and chemical changes were summarized. This will offer the reference for further research work catalpol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Catalpol, ≥96% (HPLC)