Merck
  • New insights into metabolic signaling and cell survival: the role of beta-O-linkage of N-acetylglucosamine.
New insights into metabolic signaling and cell survival: the role of beta-O-linkage of N-acetylglucosamine.
The Journal of pharmacology and experimental therapeutics (2008-09-05)
Gladys A Ngoh, Steven P Jones
ZUSAMMENFASSUNG
The involvement of glucose in fundamental metabolic pathways represents a core element of biology. Late in the 20th century, a unique glucose-derived signal was discovered, which appeared to be involved in a variety of cellular processes, including mitosis, transcription, insulin signaling, stress responses, and potentially, Alzheimer's disease, and diabetes. By definition, this glucose-fed signaling system was a post-translational modification to proteins. However, unlike classical cotranslational N-glycosylation occurring in the endoplasmic reticulum and Golgi apparatus, this process occurs elsewhere throughout the cell in a highly dynamic fashion, similar to the quintessential post-translational modification, phosphorylation. This more recently described post-translational modification, the beta-O-linkage of N-acetylglucosamine (i.e., O-GlcNAc) to nucleocytoplasmic proteins, represents an under-investigated area of biology. This signaling system operates in all of the tissues examined and seems to have persisted throughout all multicellular eukaryotes. Thus, it comes with little surprise that O-GlcNAc signaling is an integral system and viable target for biomedical investigation. This system may be a boundless source for insight into a variety of diseases and yield numerous opportunities for drug design. This Perspective will address recent insights into O-GlcNAc signaling in the cardiovascular system as a paradigm for its involvement in other biological systems.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
N-Acetyl-D-Glucosamin, ≥99% (HPLC)
Sigma-Aldrich
N-Acetyl-D-Glucosamin, suitable for cell culture, BioReagent
Sigma-Aldrich
N-Acetyl-D-Glucosamin, ≥95% (HPLC)
Supelco
N-Acetylglucosamin, Pharmaceutical Secondary Standard; Certified Reference Material