Merck
Fragrance material review on (+)-(R)-citronellol.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2008-07-22)
A Lapczynski, C S Letizia, A M Api
ZUSAMMENFASSUNG
A toxicologic and dermatologic review of (+)-(R)-citronellol when used as a fragrance ingredient is presented.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
β-Citronellol, 95%
Sigma-Aldrich
Citronellol, ≥95%, FCC, FG
Sigma-Aldrich
Citronellol, mixture of isomers, natural, ≥95%, FG
Supelco
(±)-β-Citronellol, analytical standard