Merck
  • Molecular basis of methanol-inducible gene expression and its application in the methylotrophic yeast Candida boidinii.
Molecular basis of methanol-inducible gene expression and its application in the methylotrophic yeast Candida boidinii.
Bioscience, biotechnology, and biochemistry (2009-04-09)
Hiroya Yurimoto
ZUSAMMENFASSUNG
Methanol is a promising feedstock for biotechnological and chemical processes as well as a primary source of energy to replace coal and petroleum. Methylotrophic yeasts, that can utilize methanol as the sole source of carbon and energy, have been studied intensively in terms of both physiological activities and potential applications. During growth on methanol, the enzymes involved in methanol metabolism are massively produced in these yeasts, indicating that the gene promoters of these enzymes are strong methanol-inducible promoters. Using these promoters, high-level heterologous gene expression systems have been developed in several methylotrophic yeast strains, such as Pichia pastoris, Hansenula polymorpha, and Candida boidinii. To achieve efficient industrial use of methanol and efficient protein production by methylotrophic yeasts, it is important to elucidate the molecular basis of methanol-inducible gene expression in these yeasts. This review describes recent advances in understanding of the regulation of methanol-inducible gene expression and the molecular mechanism of transcriptional activation in the methylotrophic yeast C. boidinii. Application of this gained knowledge led to successful production of useful enzymes in this yeast, which is also reviewed.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Methanol, anhydrous, 99.8%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
USP
Methylalkohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Supelco
Methanol, analytical standard
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol -Lösung, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.