Merck
Lumican, a small leucine-rich proteoglycan.
IUBMB life (2008-10-25)
Dragana Nikitovic, Paulos Katonis, Aristidis Tsatsakis, Nikos K Karamanos, George N Tzanakakis
ZUSAMMENFASSUNG
Lumican belongs to the family of small leucine-rich repeat proteoglycans. Recent studies have shown that lumican participates in the maintenance of tissue homeostasis and modulates cellular functions including cell proliferation, migration, and differentiation. The expression of lumican has been correlated to the growth and metastasis of various malignancies; however, its exact role in tumorogenesis remains elusive. This review focuses upon the role of lumican in cell biology, providing insights into molecular mechanisms that lumican likely utilizes to control processes relevant to tumorogenesis.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Leucin, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Leucin, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Leucin, BioUltra, ≥99.5% (NT)
Supelco
L-Leucin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Leucin, 99%, FG
Leucin, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Leucin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland