Merck
Calcitriol (Vectical) for mild to moderate plaque psoriasis.
The Medical letter on drugs and therapeutics (2009-09-10)
ZUSAMMENFASSUNG
The FDA has approved calcitriol ointment (Vectical--Galderma), a vitamin D analog, for topical treatment of mild-to-moderate plaque psoriasis in adults >or = 18 years old. Ointments are generally considered more potent than creams or solutions.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥99% (HPLC)
Sigma-Aldrich
1α,25-Dihydroxyvitamin D3, ≥97.0% (HPLC)
Supelco
1α,25-Dihydroxyvitamin D3 -Lösung, 5 μg/mL in ethanol, ampule of 1 mL, certified reference material, Cerilliant®
Calcitriol, European Pharmacopoeia (EP) Reference Standard