Merck
L-arginine signalling potential in the brain: the peripheral gets central.
Recent patents on CNS drug discovery (2009-06-13)
Claudiana Lameu, Antonio C M de Camargo, Marcella Faria
ZUSAMMENFASSUNG
L-arginine is a basic amino acid that has versatile metabolic roles, being involved in the generation of a wide range of biologically active intermediates such as nitric oxide (NO), polyamines, creatine and L-amino acids [1]. Because the levels of L-arginine reflect a metabolic crossroads, the mechanisms of its synthesis and degradation in peripheral tissues are very well described. However, there is an increasing amount of data also implicating this amino acid as a mediator of central nervous system activities and those are not yet fully understood. Here we shall summarize the tissue-specific pathways controlling L-arginine intracellular and blood levels and also the emerging evidence pointing to a role of this metabolite in the central regulation of diverse physiological processes, as blood pressure control and inflammatory response. As a conclusion we shall discuss the advantages of targeting L-arginine metabolism over other NO donors, as a general strategy to correct NO deficiency related diseases and discuss a few new patents recently deposited which take into account the rational perspective outlined in the present review.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
Arginin, European Pharmacopoeia (EP) Reference Standard