Merck
  • Allopurinol, an inhibitor of uric acid synthesis--can it be used for the treatment of metabolic syndrome and related disorders?
Allopurinol, an inhibitor of uric acid synthesis--can it be used for the treatment of metabolic syndrome and related disorders?
Drugs of today (Barcelona, Spain : 1998) (2009-07-09)
Iwao Suzuki, Takayuki Yamauchi, Masahiro Onuma, Shigeo Nozaki
ZUSAMMENFASSUNG
Allopurinol is an inhibitor of xanthine oxidoreductase (XOR) and inhibits the generation of uric acid (UA) as the final product of purine catabolism, as well as the resulting generation of superoxide (O2(-)), in humans. Elevation of the serum UA (SUA) level, referred to as hyperuricemia (HU), eventually leads to gout and allopurinol has been used for the treatment of HU and gout. Studies have revealed the role of elevated SUA levels and the associated oxidative stress (OS) in a broad spectrum of pathological conditions and it is anticipated that these findings would also expand the use of allopurinol as a therapeutic drug. This article presents a review of reports, mainly of recent studies, on the efficacy of allopurinol in various diseases and explores novel potential uses of the drug. Important novel and potential uses of great interest include metabolic syndrome (MetS) and related disorders, chronic kidney disease (CKD), nonalcoholic fatty liver disease (NAFLD) and nonalcoholic steatohepatitis (NASH). Ischemia-reperfusion injury and mucositis, encountered as adverse effects of cancer treatment, have also been under investigation as potential targets for allopurinol.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Allopurinol, xanthine oxidase inhibitor
Supelco
Allopurinol, Pharmaceutical Secondary Standard; Certified Reference Material
Allopurinol, European Pharmacopoeia (EP) Reference Standard