Merck
Ethinylestradiol/dienogest in oral contraception.
Drugs (2010-04-17)
Ezequiel F Pérez-Campos
ZUSAMMENFASSUNG
The low-dose combined oral contraceptive of ethinylestradiol 30 microg and dienogest 2 mg was launched in Germany in 1995, and is now the most commonly prescribed oral contraceptive in this country. Dienogest is a novel 19-nortestosterone-derived progestin with a unique pharmacokinetic and pharmacological profile, including antiandrogenic properties. Clinical studies have demonstrated that ethinylestradiol/dienogest is a reliable ovulation inhibitor with high contraceptive efficacy that is comparable with other combined oral contraceptives. It also provides effective cycle control, with reduced intensity and duration of menstrual bleeding, and improves dysmenorrhoea. The combination of ethinylestradiol and dienogest reduces serum androgen levels, and increases the levels of thyroid hormones; however, although thyroid hormone levels increase, there is no increased activity due to increases in transporter protein. Like other low-dose oral contraceptives, ethinylestradiol/dienogest has only minor influences on lipid and carbohydrate metabolism, adrenal hormones and blood pressure parameters, and appears to have a balanced effect on the haemostatic system. Ethinylestradiol/dienogest also has beneficial effects on hair and skin; a number of studies have reported decreased hair and skin greasiness, and improvements in acne vulgaris following treatment with ethinylestradiol/dienogest. After discontinuation of ethinylestradiol/dienogest, there may be a small delay in conception during the first three cycles, but there is no subsequent impairment of fertility. Furthermore, the duration of use of ethinylestradiol/dienogest does not seem to influence the rate of conception or time to conception. Ethinylestradiol/dienogest is well tolerated; adverse reactions associated with treatment include breast pain, headache and nausea/vomiting. These adverse reactions are rare and decrease in incidence over time.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
17α-Ethinylestradiol, ≥98%
Supelco
Ethinylestradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethinylestradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Ethinylestradiol für die Systemeignung, European Pharmacopoeia (EP) Reference Standard