Merck
Industrial-grade talc exposure and the risk of mesothelioma.
Critical reviews in toxicology (2010-03-17)
Bertram Price
ZUSAMMENFASSUNG
Industrial-grade talc deposits are complex mixtures of mineral particles and may vary substantially in composition across small geographical areas. Typical industrial-grade talc includes amphibole cleavage fragments, platy talc, serpentine minerals, talc in fibrous form, and a minor presence of transitional fibers. Industrial-grade talc was erroneously determined to be an asbestos-containing material due to an unintended consequence of Occupational Health and Safety Administration's (OSHA's) method for measuring airborne asbestos mandated in 1972. This error was repeated, most notably, by the National Institute for Occupational Safety and Health (NIOSH) in, 1980 for talc mined in northern New York State (NYS) by RT Vanderbilt Company (RTV). Subsequent exposure studies of northern NYS talc conducted through the, 1980s and one study published after, 2000 relied on the conclusion that talc was an asbestos-containing material to infer a causal relationship between talc and mesothelioma. The present review included (1) publications concerning talc's cancer-causing potential issued by organizations concerned with occupational and public health; (2) talc exposure studies and animal and cellular studies of RTV talc; (3) mesothelioma rates in northern NYS; and (4) mesothelioma mortality among RTV mining employees. The review indicated that failure to correctly identify the mineral characteristics of talc resulted in misleading reports concerning the carcinogenic potential of talc. However, the collective data from animal and cellular studies, mesothelioma rates in northern NYS, exposure studies, and a mortality analysis of RTV mining employees do not support a causal relationship between RTV talc and mesothelioma. This conclusion is applicable to all mineral components in RTV talc and to other industrial-grade talcs and mineral aggregates with the same components.