Merck
Targeting the L-arginine-nitric oxide pathway for cancer treatment.
Current pharmaceutical design (2010-03-20)
Qingyong Ma, Zheng Wang, Min Zhang, Hengtong Hu, Junhui Li, Dong Zhang, Kun Guo, Huanchen Sha
ZUSAMMENFASSUNG
The action of L-arginine is mainly dependent on its end-product, nitric oxide (NO). The L-arginine/NO pathway has been confirmed to play an important role in tumor development. Recent findings indicate that NO derived from L-arginine could influence angiogenesis factors, vascular permeability, perivascular-cell recruitment and vessel remodeling and maturation. Additionally, the L-arginine/NO pathway could activate a broad array of genes that are functionally involved in proliferation, metastasis and apoptosis. Interestingly, this pathway plays roles in both tumorigensis and tumor killing. The role of the L-arginine/NO pathway in tumor therapy has been well-studied. Members of this pathway have been reported to be promising therapeutic molecules in tumor therapy. This review article summarizes research data on the roles of the L-arginine/NO pathway in cancer biology and cancer therapy.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
L-Arginin, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Arginin, reagent grade, ≥98%
Sigma-Aldrich
L-Arginin, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Arginin, 99%, FCC, FG
SAFC
L-Arginin
Supelco
L-Arginin, Pharmaceutical Secondary Standard; Certified Reference Material
Arginin, European Pharmacopoeia (EP) Reference Standard