Merck
Dextran-doxorubicin/chitosan nanoparticles for solid tumor therapy.
Wiley interdisciplinary reviews. Nanomedicine and nanobiotechnology (2010-01-06)
Savita Bisht, Amarnath Maitra
ZUSAMMENFASSUNG
Chemotherapy is a major therapeutic approach for the treatment of localized and metastasized cancers. Whereas potent chemotherapeutic agents seem promising in the test tube, clinical trials often fail due to unfavorable pharmacokinetics, poor delivery, low local concentrations, and limited accumulation in the target cell. The pathophysiology of the tumor vasculature and stromal compartment presents a major obstacle to effective delivery of agents to solid tumors. Poor perfusion of the tumor, arterio-venous shunting, necrotic and hypoxic areas, as well as a high interstitial fluid pressure work against favorable drug uptake. Thus, targeted drug delivery using long-circulating particulate drug carriers such as hydrogels of controlled size (<100 nm diameter) holds immense potential to improve the treatment of cancer by selectively providing therapeutically effective drug concentrations at the tumor site [through enhanced permeability and retention (EPR) effect] while reducing undesirable side effects. This review focuses on the progress of targeted delivery of nanoparticulated anticancer drug such as doxorubicin chemically conjugated with dextran and encapsulated in chitosan nanoparticles to solid tumor with reduced side effect of drug. Regulated particle size and long circulation of these hydrogel nanoparticles in blood help them accumulate in tumor tissue through EPR effect as evident from the significant regression of the tumor volume. The cardiotoxicity of doxorubicin can be minimized by coupling the drug with dextran and encapsulating it in chitosan nanoparticles.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~70,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~40,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 60,000-76,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr ~100,000
Supelco
Dextran, analytical standard, 25,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 150,000
Supelco
Dextran, analytical standard, 5,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 670,000
Supelco
Dextran, analytical standard, suitable for gel permeation chromatography (GPC), Set Mp 1,000-400,000
Supelco
Dextran, analytical standard, 670,000
Supelco
Dextran, analytical standard, 150,000
Supelco
Dextran, analytical standard, 1,000
Supelco
Dextran, analytical standard, 270,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran, analytical standard, 12,000
Sigma-Aldrich
Dextran aus Leuconostoc spp., Mr 15,000-25,000
Sigma-Aldrich
Dextran -Lösung aus Leuconostoc mesenteroides, 20 % (w/w) (Autoclaved)
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, Mr ~60,000
Supelco
Dextran, analytical standard, 410,000
Sigma-Aldrich
Dextran, enzymatic synth.
Supelco
Dextran, analytical standard, 50,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, suitable for gel permeation chromatography (GPC), Mw 5,000
Supelco
Dextran, analytical standard, 80,000
Supelco
Dextran aus Leuconostoc mesenteroides, analytical standard, Mw 1,000
Sigma-Aldrich
Dextran aus Leuconostoc mesenteroides, average mol wt 48,000-90,000