Merck
  • [Recent advances in bacterial biodegradation of naphthalene, phenanthrene by bacteria: a review].
[Recent advances in bacterial biodegradation of naphthalene, phenanthrene by bacteria: a review].
Sheng wu gong cheng xue bao = Chinese journal of biotechnology (2010-09-08)
Dan Zhang, Zhaoge Li, Xinguang Bao, Jiangbo Li, Haihua Liang, Kangmin Duan, Lixin Shen
ZUSAMMENFASSUNG
Polycyclic aromatic hydrocarbons (PAHs) are toxic pollutants that exist extensively in the environment. Microbial degradation is the main pathway of PAHs eradication in natural environment and therefore is of importance to investigate. Advancement has been made in recent years regarding the PAHs molecular degradation mechanisms in bacteria. In this review, we summarized some of the research progresses in microbial PAHs biodegradation pathways (including salicylate pathway and protocatechuate pathway), key enzymes (nah-like, phn, phd, nid and nag) and genes involved. Emphasis was given on naphthalene and phenanthrene which were often used as the representatives of PAHs. It is likely that the new information will promote further research and applications of microbial PAHs biodegradation technology.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Naphthalin, 99%
Sigma-Aldrich
Phenanthren, 98%
Supelco
Naphthalin, analytical standard
Sigma-Aldrich
Naphthalin, suitable for scintillation, ≥99%
Supelco
Schmelzpunktstandard 79-81 °C, analytical standard
Supelco
Naphthalin, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Naphthalin -Lösung, certified reference material, 5000 μg/mL in methanol
Sigma-Aldrich
Phenanthren, sublimed grade, ≥99.5%
Supelco
Phenanthren, analytical standard, for environmental analysis
Supelco
Phenanthren, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Naphthalin, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland