Merck
The health promoting properties of the conjugated isomers of α-linolenic acid.
Lipids (2010-12-17)
Alan A Hennessy, R Paul Ross, Rosaleen Devery, Catherine Stanton
ZUSAMMENFASSUNG
The bioactive properties of the conjugated linoleic acid (CLA) isomers have long been recognised and are the subject of a number of excellent reviews. However, despite this prominence the CLA isomers are not the only group of naturally occurring dietary conjugated fatty acids which have shown potent bioactivity. In a large number of in vitro and in vivo studies, conjugated α-linolenic acid (CLNA) isomers have displayed potent anti-inflammatory, immunomodulatory, anti-obese and anti-carcinogenic activity, along with the ability to improve biomarkers of cardio-vascular health. CLNA isomers are naturally present in high concentrations in a large variety of seed oils but can also be produced in vitro by strains of lactobacilli and bifidobactena through the activity of the enzyme linoleic acid isomerase on α-linolenic acid. In this review, we will address the possible therapeutic roles that CLNA may play in a number of conditions afflicting Western society and the mechanisms through which this activity is mediated.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Linolensäure, ≥99%
Supelco
Linolensäure, analytical standard
Sigma-Aldrich
Linolensäure, ~70% (GC)