Merck
  • Reperfusion of chronic tissue ischemia: nitrite and dipyridamole regulation of innate immune responses.
Reperfusion of chronic tissue ischemia: nitrite and dipyridamole regulation of innate immune responses.
Annals of the New York Academy of Sciences (2010-10-20)
Christopher B Pattillo, Kai Fang, Justin Terracciano, Christopher G Kevil
ZUSAMMENFASSUNG
Chronic and intermittent ischemic vascular disorders represent a burgeoning clinical challenge. Previous studies have focused on the idea that therapeutic angiogenesis strategies could alleviate tissue ischemia; however, it is now appreciated that vascular disease is not simply limited to vascular wall cells but also influenced by simultaneously occurring inflammatory responses. Our laboratory has discovered that pharmacological treatment of permanent tissue ischemia with dipyridamole significantly augments ischemic tissue reperfusion, angiogenesis, and arteriogenesis over time. We have found that the beneficial effects of dipyridamole therapy are due to its ability to increase tissue nitric oxide bioavailability that corrects tissue redox imbalance. Importantly, we have also discovered that dipyridamole treatment invoking nitric oxide (NO) production significantly downregulates various innate immune response genes during chronic ischemic tissue injury. These findings demonstrate that dipyridamole-induced production of nitrite/NO significantly decreases inflammatory responses while increasing vascular growth in ischemic tissues.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Dipyridamol, ≥98% (HPLC)
Dipyridamol für die Peakidentifizierung, European Pharmacopoeia (EP) Reference Standard
Dipyridamol, European Pharmacopoeia (EP) Reference Standard