Merck
Drospirenone/ethinylestradiol: a review on efficacy and noncontraceptive benefits.
Women's health (London, England) (2010-12-24)
Rogério Bonassi Machado, Luciano de Melo Pompei, Arícia Galvão Giribela, Cassiana Galvão Giribela
ZUSAMMENFASSUNG
Considerable advances have been made in the field of combined oral contraceptives since their introduction 50 years ago. This ongoing evolution has been focused on reducing the estrogen dose and synthesizing new progestogens. A recently launched combination of ethinylestradiol 20 µg and drospirenone 3 mg demonstrates the advantages of a lower estrogen dose with the antimineralocorticoid activity of drospirenone that is responsible for the drug's significant antiandrogenic and antimineralocorticoid effects, reflected clinically in lower rates of adverse events including less fluid retention. In addition to the drug's contraceptive efficacy, its effects on the skin and other classic noncontraceptive benefits, the ethinylestradiol 20 µg and drospirenone 3 mg combination has demonstrated highly satisfactory results in women with premenstrual dysphoric syndrome.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
17α-Ethinylestradiol, ≥98%
Supelco
Ethinylestradiol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Ethinylestradiol, United States Pharmacopeia (USP) Reference Standard
Ethinylestradiol, European Pharmacopoeia (EP) Reference Standard
Supelco
17α-Ethynylestradiol solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Drospirenon, ≥98% (HPLC)
Ethinylestradiol für die Systemeignung, European Pharmacopoeia (EP) Reference Standard
Drospirenon, European Pharmacopoeia (EP) Reference Standard