Merck
Pravastatin versus simvastatin for prevention of contrast-induced nephropathy.
Journal of cardiovascular pharmacology and therapeutics (2011-03-10)
Monica A Muñoz, Pamela R Maxwell, Kay Green, Darrel W Hughes, Robert L Talbert
ZUSAMMENFASSUNG
Contrast-induced nephropathy (CIN) is associated with long-term morbidity, mortality, and increased health care costs. It has been suggested that statins have pleiotropic effects countering inflammatory and oxidative stress involved in CIN. Several studies support this theory; however, previously published studies have not evaluated the potential differences between statins in reducing the incidence of CIN. The purpose of this retrospective, single-center trial was to compare the incidence of CIN in patients receiving simvastatin or pravastatin therapy undergoing percutaneous coronary intervention (PCI). A total of 261 patients were included (145 received simvastatin and 116 received pravastatin) with the majority undergoing elective PCI. The population was predominantly male (65%), Hispanic (65%), and diabetic (62%), with a mean age of 59 years and a low-density lipoprotein (LDL) of 85 mg/dL. No significant differences were found between groups for risk factors or prophylactic strategies (eg, hydration). Contrast-induced nephropathy occurred in 26 patients (17.9%) in the simvastatin group versus 10 (8.6%) in the pravastatin group (P < .05). No patients required dialysis as a result of contrast administration. Acute kidney injury (AKI) occurred in 21 patients (14.5%) in the simvastatin group compared to 8 (6.9%) in the pravastatin group (P < .05). In multivariate analysis, the difference between statins remained an independent predictor for the development of CIN. In conclusion, patients on pravastatin had a significantly lower incidence of CIN compared to patients on simvastatin.
MATERIALIEN
Produktnummer
Marke
Produktbeschreibung
Sigma-Aldrich
Simvastatin, ≥97% (HPLC), solid
Supelco
Simvastatin, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Simvastatin, United States Pharmacopeia (USP) Reference Standard
Simvastatin, European Pharmacopoeia (EP) Reference Standard
Supelco
Simvastatin, analytical standard